Dithallium selenide ≥99.9%

Supplier: BeanTown Chemical

Synonyms: Thallium(I) selenide

214910-1G 214910-5G
BT214910-1GEA 57 USD
BT214910-1G BT214910-5G
Dithallium selenide ≥99.9%
Dithallium selenide

CAS: 15572-25-5; EC No: 239-627-1; MDL No: MFCD00049603 UN No: UN1707; Haz Class: 6.1; Packing Group: II 10mm & Down Lump; Linear Formula: Tl2Se; MW: 487.72 Melting Point: 340° Density (g/mL): 9.05


  • 10mm & Down Lump

Formula: SeTl₂
MW: 487.73 g/mol
Melting Pt: 340 °C
Density: 9.05 g/mL
MDL Number: MFCD00049603
CAS Number: 15572-25-5
EINECS: 239-627-1
UN: 1707
ADR: 6.1 II
Merck Index: 12,09403

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR