Methyl dichloroacetate ≥99%
Supplier: BeanTown Chemical
214840-100G
214840-500G
BT214840-100GEA
42.57
USD
BT214840-100G
BT214840-500G
Methyl dichloroacetate ≥99%
Methyl dichloroacetate
CAS: 116-54-1; EC No: 204-146-8; MDL No: MFCD00000843; RTECS: AG6625000
UN No: UN2299; Haz Class: 6.1; Packing Group: III
Liquid; Linear Formula: Cl2CHCOOCH3; Molecular Formula: C3H4Cl2O2; MW: 142.97
Melting Point: -52°; Boiling Point: 143°; Flash point: 64°C (147°F)
Density (g/mL): 1.381; Refractive Index: 1.442
- RTECS: AG6625000
- Refractive index: 1.442
- Liquid
Formula:
C₃H₄Cl₂O₂ MW: 142.97 g/mol Boiling Pt: 143 °C Melting Pt: –52 °C Density: 1.381 g/mL Flash Pt: 64 °C (147 °F) |
MDL Number:
MFCD00000843 CAS Number: 116-54-1 EINECS: 204-146-8 UN: 2299 ADR: 6.1,III |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...