4'-Isobutylacetophenone ≥97%

Supplier: BeanTown Chemical

Synonyms: p-Isobutylacetophenone

214820-250G 214820-50G
BT214820-250GEA 456 USD
BT214820-250G BT214820-50G
4'-Isobutylacetophenone ≥97%
4'-Isobutylacetophenone

CAS: 38861-78-8; EC No: 254-159-8; MDL No: MFCD00027393 UN No: UN1224; Haz Class: 3; Packing Group: III Liquid; Molecular Formula: C12H16O; MW: 176.27 Flash point: 54°C (129°F) Density (g/mL): 0.952; Refractive Index: 1.518


  • Refractive index: 1.518
  • Liquid

Formula: C₁₂H₁₆O
MW: 176.26 g/mol
Density: 0.952 g/mL
Flash Pt: 54 °C (129 °F)
MDL Number: MFCD00027393
CAS Number: 38861-78-8
EINECS: 254-159-8
UN: 1224
ADR: 3,III

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR