Chlorodimethyl(pentafluorophenyl)silane (flophemesyl chloride) ≥96%

Supplier: BeanTown Chemical

Synonyms: (Chloro)(perfluorophenyl)dimethylsilane

214780-1G 214780-5G
BT214780-1GEA 39 USD
BT214780-1G BT214780-5G
Chlorodimethyl(pentafluorophenyl)silane (flophemesyl chloride) ≥96%
Chlorodimethyl(pentafluorophenyl)silane (flophemesyl chloride)

CAS: 20082-71-7; EC No: 243-506-9; MDL No: MFCD00000500 UN No: UN2987; Haz Class: 8; Packing Group: II Liquid; Molecular Formula: C8H6ClF5Si; MW: 260.66 Boiling Point: 88-90°/10 mmHg; Flash point: 95°C (203°F) Density (g/mL): 1.384; Refractive Index: 1.448 Moisture Sensitive


  • Refractive index: 1.448
  • Moisture Sensitive
  • Liquid

Formula: C₈H₆ClF₅Si
MW: 260.67 g/mol
Boiling Pt: 88…90 °C/10 mmHg
Density: 1.384 g/mL
Flash Pt: 95 °C (203 °F)
MDL Number: MFCD00000500
CAS Number: 20082-71-7
EINECS: 243-506-9
UN: 2987
ADR: 8,II

Order Now

Specification Test Results

Assay 96%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR