α'-Bromo-α,α,α-trifluoro-o-xylene ≥97%

Supplier: BeanTown Chemical

214775-1G 214775-25G 214775-5G
BT214775-1GEA 23 USD
BT214775-1G BT214775-25G BT214775-5G
α'-Bromo-α,α,α-trifluoro-o-xylene ≥97%
α'-Bromo-α,α,α-trifluoro-o-xylene

CAS: 395-44-8; EC No: 206-901-7; MDL No: MFCD00013558 UN No: UN3261; Haz Class: 8; Packing Group: II Liquid/Low Melting Solid; Molecular Formula: C9H7F3; MW: 239.03 Boiling Point: 72°/7.5 mmHg; Flash point: 83°C (181°F) Density (g/mL): 1.571; Refractive Index: 1.494


  • Refractive index: 1.494
  • Liquid/Low Melting Solid

Formula: C₈H₆BrF₃
Boiling Pt: 72 °C/7.5 mmHg
Density: 1.571 g/mL
Flash Pt: 83 °C (181 °F)
MDL Number: MFCD00013558
CAS Number: 395-44-8
EINECS: 206-901-7
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR