Tetraamminepalladium(II) nitrate (≤5.0% Pd) ≥99.9% (trace metals basis)

Supplier: BeanTown Chemical

214750-C10G 214750-C2G
BT214750-C10GEA 1833.11 USD
BT214750-C10G BT214750-C2G
Tetraamminepalladium(II) nitrate (≤5.0% Pd) ≥99.9% (trace metals basis)
Tetraamminepalladium(II) nitrate

CAS: 13601-08-6; EC No: 237-078-2; MDL No: MFCD00011591 UN No: UN3218; Haz Class: 5.1; Packing Group: III Liquid; Linear Formula: Pd(NH3)4(NO3)2; Molecular Formula: H12N6O6Pd; MW: 298.63


  • Liquid

Formula: H₁₂N₆O₆Pd
MW: 298.552 g/mol
MDL Number: MFCD00011591
CAS Number: 13601-08-6
EINECS: 237-078-2
ADR: 5.1,III

Order Now

Specification Test Results

Assay 99.9%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR