1-Cyclohexyl-2-pyrrolidone ≥99%
Supplier: BeanTown Chemical
214740-250G
214740-50G
BT214740-250GEA
39
USD
BT214740-250G
BT214740-50G
1-Cyclohexyl-2-pyrrolidone ≥99%
1-Cyclohexyl-2-pyrrolidone
CAS: 6837-24-7; EC No: 229-919-7; MDL No: MFCD00003191; RTECS: UY5748450
UN No: UN2810; Haz Class: 6.1; Packing Group: II
Liquid; Molecular Formula: C10H17NO; MW: 167.25
Boiling Point: 154°/7 mmHg; Flash point: 141°C (286°F)
Density (g/mL): 1.007; Refractive Index: 1.499
- RTECS: UY5748450
- Refractive index: 1.499
- Liquid
Formula:
C₁₀H₁₇NO MW: 167.25 g/mol Boiling Pt: 154 °C/7 mmHg Density: 1.007 g/mL Flash Pt: 141 °C (286 °F) |
MDL Number:
MFCD00003191 CAS Number: 6837-24-7 EINECS: 229-919-7 ADR: 6.1,II |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...