2-Amino-6-(trifluoromethoxy)benzothiazole ≥98%

Supplier: BeanTown Chemical

Synonyms: 6-(Trifluoromethoxy)benzo[d]thiazol-2-amine, Riluzole

214735-25G 214735-5G
BT214735-25GEA 204 USD
BT214735-25G BT214735-5G
2-Amino-6-(trifluoromethoxy)benzothiazole ≥98%
2-Amino-6-(trifluoromethoxy)benzothiazole

CAS: 1744-22-5; MDL No: MFCD00210213; RTECS: DL2830000 UN No: UN2811; Haz Class: 6.1; Packing Group: II Solid; Molecular Formula: C8H5ClF3N2OS; MW: 234.20 Melting Point: 116-118°


  • RTECS: DL2830000
  • Solid

Formula: C₈H₅F₃N₂OS
Melting Pt: 116…118 °C
MDL Number: MFCD00210213
CAS Number: 1744-22-5
ADR: 6.1,II

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR