Dicyclohexylmethane-4,4'-diisocyanate (mixture of isomers) ≥90%, Technical Grade
Supplier: BeanTown Chemical
CAS: 5124-30-1; EC No: 225-863-2; MDL No: MFCD00015405; RTECS: NQ9250000
UN No: UN2206; Haz Class: 6.1; Packing Group: III
Liquid; Molecular Formula: C15H22N2O2; MW: 262.37
Melting Point: 26°; Flash point: 200°C (392°F)
Density (g/mL): 1.066; Refractive Index: 1.497
- RTECS: NQ9250000
- Refractive index: 1.497
- Liquid
Formula:
C₁₅H₂₂N₂O₂ Melting Pt: 26 °C Density: 1.066 g/mL Flash Pt: 200 °C (392 °F) |
MDL Number:
MFCD00015405 CAS Number: 5124-30-1 EINECS: 225-863-2 ADR: 6.1,III |
Specification Test Results
Assay | 90% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...