Diisopropyl cyanomethylphosphonate ≥97%
Supplier: BeanTown Chemical
214680-25G
214680-5G
BT214680-25GEA
338
USD
BT214680-25G
BT214680-5G
Diisopropyl cyanomethylphosphonate ≥97%
Diisopropyl cyanomethylphosphonate
CAS: 21658-95-7; EC No: 244-511-9; MDL No: MFCD00009812
UN No: UN2927; Haz Class: 6.1 (8); Packing Group: II
Liquid; Molecular Formula: C8H16NO3P ; MW: 205.19
Boiling Point: 108-111°/0.08 mmHg
Density (g/mL): 1.039
Moisture Sensitive
- Moisture Sensitive
- Liquid
Formula:
C₈H₁₆NO₃P MW: 205.194 g/mol Boiling Pt: 108…111 °C/0.08 mmHg Density: 1.039 g/mL |
MDL Number:
MFCD00009812 CAS Number: 21658-95-7 EINECS: 244-511-9 ADR: 6.1 (8) II |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...