Cobalt(II) chloride hexahydrate ≥98%

Supplier: BeanTown Chemical

Synonyms: Cobalt Chloride, Hexahydrate, Cobalt dichloride hexahydrate, Cobalt Chloride, 6-Hydrate, Cobaltous chloride hexahydrate

214635-1KG 214635-250G 214635-50G
BT214635-1KGEA 210.73 USD
BT214635-1KG BT214635-250G BT214635-50G
Cobalt(II) chloride hexahydrate ≥98%
Cobalt(II) chloride hexahydrate

CAS: 7791-13-1; EC No: 231-589-4; MDL No: MFCD00149652; RTECS: GG0200000 UN No: UN3077; Haz Class: 9; Packing Group: III Solid; Linear Formula: CoCl2·6H2O; MW: 237.93 Hygroscopic


  • RTECS: GG0200000
  • Hygroscopic
  • Solid

Formula: CoCl₂·6H₂O
MW: 237.93 g/mol
Boiling Pt: 110
Melting Pt: 56
MDL Number: MFCD00149652
CAS Number: 7791-13-1
EINECS: 231-589-4
UN: 3077
ADR: 9 III
Merck Index: 13,02462

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR