Mercury(II) telluride ≥99.999%
Supplier: BeanTown Chemical
Synonyms:
Mercurous telluride
214610-1G
214610-5G
BT214610-1GEA
63
USD
BT214610-1G
BT214610-5G
Mercury(II) telluride ≥99.999%
Mercury(II) telluride
CAS: 12068-90-5; EC No: 235-108-9; MDL No: MFCD00016133
UN No: UN2025; Haz Class: 6.1; Packing Group: II
3-12mm Fused Lump; Molecular Formula: HgTe; MW: 328.19
Melting Point: 673°
Density (g/mL): 8.63
- 3-12mm Fused Lump
Formula:
HgTe MW: 328.19 Melting Pt: 550 Density: 8.63 g/mL |
MDL Number:
MFCD00016133 CAS Number: 12068-90-5 EINECS: 235-108-9 UN: 2025 ADR: 6.1 II |
Specification Test Results
Assay | 99.999% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...