2,2,2-Trifluoroethyl acetate ≥97%
Supplier: BeanTown Chemical
214565-100G
214565-25G
BT214565-100GEA
213
USD
BT214565-100G
BT214565-25G
2,2,2-Trifluoroethyl acetate ≥97%
2,2,2-Trifluoroethyl acetate
Formula:
C₄H₅F₃O₂ Density: 1.258 g/mL Flash Pt: 11 °C (51 °F) |
MDL Number:
MFCD00198018 CAS Number: 406-95-1 UN: 3272 ADR: 3,II |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...