Phenyltrimethylammonium iodide ≥99%

Supplier: BeanTown Chemical

Synonyms: N,N,N-Trimethylanilinium iodide, Trimethylphenylammonium iodide

214535-100G 214535-25G 214535-500G
BT214535-100GEA 63 USD
BT214535-100G BT214535-25G BT214535-500G
Phenyltrimethylammonium iodide ≥99%
Phenyltrimethylammonium iodide

CAS: 98-04-4; EC No: 202-630-3; MDL No: MFCD00011791; RTECS: BT2450000 UN No: UN2811; Haz Class: 6.1; Packing Group: III Powder; Linear Formula: C6H5N(CH3)3I; Molecular Formula: C9H14IN; MW: 263.12 Melting Point: 229° (decomposes) Hygroscopic , Light Sensitive


  • RTECS: BT2450000
  • Hygroscopic, Light Sensitive
  • Powder

Formula: C₉H₁₄IN
MW: 263.12 g/mol
Melting Pt: 229 °C (dec.)
MDL Number: MFCD00011791
CAS Number: 98-04-4
EINECS: 202-630-3
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR