Trityl chloride ≥98%

Supplier: BeanTown Chemical

Synonyms: Trt-Cl, Chlorotriphenylmethane, Triphenylmethyl chloride, (Chloromethanetriyl)tribenzene, Triphenylchloromethane

214520-100G 214520-25G 214520-500G
BT214520-100GEA 53.77 USD
BT214520-100G BT214520-25G BT214520-500G
Trityl chloride ≥98%
Trityl chloride

CAS: 76-83-5; EC No: 200-986-4; MDL No: MFCD00000813; RTECS: PA6450000 UN No: UN3261; Haz Class: 8; Packing Group: II Powder; Linear Formula: (C6H5)3CCl; Molecular Formula: C19H15Cl; MW: 278.78 Melting Point: 109-112° Moisture Sensitive


  • RTECS: PA6450000
  • Moisture Sensitive
  • Powder

Formula: C₁₉H₁₅Cl
MW: 278.78 g/mol
Melting Pt: 109…112 °C
MDL Number: MFCD00000813
CAS Number: 76-83-5
EINECS: 200-986-4
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR