3-Bromo-2-thiophenecarbonitrile ≥97%
Supplier: BeanTown Chemical
Synonyms:
3-Bromothiophene-2-carbonitrile
214510-1G
214510-250MG
BT214510-1GEA
262
USD
BT214510-1G
BT214510-250MG
3-Bromo-2-thiophenecarbonitrile ≥97%
3-Bromo-2-thiophenecarbonitrile
CAS: 18791-98-5; MDL No: MFCD00068173
UN No: UN3439; Haz Class: 6.1; Packing Group: III
Powder; Molecular Formula: C5H2BrNS; MW: 188.05
- Powder
Formula:
C₅H₂BrNS MW: 188.05 g/mol |
MDL Number:
MFCD00068173 CAS Number: 18791-98-5 UN: 3439 ADR: 6.1,III |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...