Benzophenone hydrazone ≥98%

Supplier: BeanTown Chemical

214385-1KG 214385-250G 214385-50G
BT214385-1KGEA 251 USD
BT214385-1KG BT214385-250G BT214385-50G
Benzophenone hydrazone ≥98%
Benzophenone hydrazone

CAS: 5350-57-2; EC No: 226-321-8; MDL No: MFCD00007624; RTECS: PC4954487 UN No: UN2811; Haz Class: 6.1; Packing Group: III Crystalline; Molecular Formula: C13H12N2 ; MW: 196.25 Melting Point: 95-98°; Boiling Point: 225-230°/55 mmHg


  • RTECS: PC4954487
  • Crystalline

Formula: C₁₃H₁₂N₂
MW: 196.25 g/mol
Boiling Pt: 225…230 °C/55 mmHg
Melting Pt: 95…98 °C
MDL Number: MFCD00007624
CAS Number: 5350-57-2
EINECS: 226-321-8
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR