1,1,2,2-Tetrachloroethane ≥98%
Supplier: BeanTown Chemical
CAS: 79-34-5; EC No: 201-197-8; MDL No: MFCD00000848; RTECS: KI8575000
UN No: UN1702; Haz Class: 6.1; Packing Group: II
Liquid; Linear Formula: Cl2CHCHCl2; MW: 167.85
Melting Point: -43°; Boiling Point: 147°
Density (g/mL): 1.596; Refractive Index: 1.494
- RTECS: KI8575000
- Refractive index: 1.494
- Liquid
Formula:
Cl₂HCCHCl₂ Boiling Pt: 147 °C Melting Pt: –43 °C Density: 1.596 g/mL |
MDL Number:
MFCD00000848 CAS Number: 79-34-5 EINECS: 201-197-8 UN: 1702 ADR: 6.1,II |
|
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...