Indium(III) trifluoromethanesulphonate ≥99%
Supplier: BeanTown Chemical
Synonyms:
Indium(III) trifluoromethanesulphonate, Indium triflate
214280-10G
214280-50G
BT214280-10GEA
129.46
USD
BT214280-10G
BT214280-50G
Indium(III) trifluoromethanesulphonate ≥99%
Indium(III) trifluoromethanesulphonate
CAS: 128008-30-0; MDL No: MFCD00144478
UN No: UN3261; Haz Class: 8; Packing Group: II
Powder; Linear Formula: In(CF3SO3)3; MW: 562.02
Hygroscopic
- Hygroscopic
- Powder
Formula:
C₃F₉InO₉S₃ MW: 562.03 |
MDL Number:
MFCD00144478 CAS Number: 128008-30-0 UN: 3261 ADR: 8 II |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...