Trifluoromethanesulfonic anhydride ≥98%
Supplier: BeanTown Chemical
Synonyms:
Triflic anhydride
214265-25G
214265-50G
214265-5G
BT214265-25GEA
143.17
USD
BT214265-25G
BT214265-50G
BT214265-5G
Trifluoromethanesulfonic anhydride ≥98%
Trifluoromethanesulphonic anhydride
CAS: 358-23-6; EC No: 206-616-8; MDL No: MFCD00000408; RTECS: PB2772000
UN No: UN3265; Haz Class: 8; Packing Group: II
Liquid; Linear Formula: (CF3SO2)2O; MW: 282.13
Melting Point: -80°; Boiling Point: 81-83°
Density (g/mL): 1.677; Refractive Index: 1.321
Moisture Sensitive
- RTECS: PB2772000
- Refractive index: 1.321
- Moisture Sensitive
- Liquid
Formula:
C₂F₆O₅S₂ MW: 282.14 g/mol Boiling Pt: 81…83 °C Melting Pt: –80 °C Density: 1.677 g/mL |
MDL Number:
MFCD00000408 CAS Number: 358-23-6 EINECS: 206-616-8 UN: 3265 ADR: 8,II |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...