Zinc 2-ethylhexanoate ≥80% (w/w) in mineral oil
Supplier: BeanTown Chemical
Synonyms:
Zinc(II) 2-ethylhexanoate
214190-100G
214190-2.5KG
214190-500G
BT214190-100GEA
23
USD
BT214190-100G
BT214190-2.5KG
BT214190-500G
Zinc 2-ethylhexanoate ≥80% (w/w) in mineral oil
Zinc 2-ethylhexanoate
CAS: 136-53-8; EC No: 205-251-1; MDL No: MFCD00053316
UN No: UN1268; Haz Class: 3; Packing Group: III
Liquid; Linear Formula: [CH3(CH2)3CH(C2H5)CO2]2Zn; Molecular Formula: C16H30O4Zn ; MW: 351.79
Density (g/mL): 1.07
- Liquid
Formula:
C₁₆H₃₀O₄Zn Density: 1.07 g/mL |
MDL Number:
MFCD00053316 CAS Number: 136-53-8 EINECS: 205-251-1 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...