Tributylamine ≥98%
Supplier: BeanTown Chemical
CAS: 102-82-9; EC No: 203-058-7; MDL No: MFCD00009431; RTECS: YA0350000
UN No: UN2542; Haz Class: 6.1; Packing Group: II
Liquid; Linear Formula: [CH3(CH2)3]3N; Molecular Formula: C12H27N; MW: 185.35
Melting Point: -70°; Boiling Point: 216°; Flash point: 63°C (145°F)
Density (g/mL): 0.778; Refractive Index: 1.428
Hygroscopic
- RTECS: YA0350000
- Refractive index: 1.428
- Hygroscopic
- Liquid
Formula:
C₁₂H₂₇N Boiling Pt: 216 °C Melting Pt: –70 °C Density: 0.778 g/mL Flash Pt: 63 °C (145 °F) |
MDL Number:
MFCD00009431 CAS Number: 102-82-9 EINECS: 203-058-7 UN: 2542 ADR: 6.1,II |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...