Copper(I) oxide ≥97%

Supplier: BeanTown Chemical

Synonyms: Cuprous oxide, Dicopper oxide

214080-2KG 214080-500G
BT214080-2KGEA 108.57 USD
BT214080-2KG BT214080-500G
Copper(I) oxide ≥97%
Copper(I) oxide

CAS: 1317-39-1; EC No: 215-270-7; MDL No: MFCD00010974; RTECS: GL8050000 UN No: UN3077; Haz Class: 9; Packing Group: III -325 Mesh Powder; Molecular Formula: Cu2O; MW: 143.08 Melting Point: 1232° Density (g/mL): 6 Air Sensitive, Moisture Sensitive


  • RTECS: GL8050000
  • Air Sensitive, Moisture Sensitive
  • -325 Mesh Powder

Formula: Cu₂O
MW: 143.09 g/mol
Melting Pt: 1232 °C
Density: 6 g/mL
MDL Number: MFCD00010974
CAS Number: 1317-39-1
EINECS: 215-270-7
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR