Cobalt(II) chloride hexahydrate 98.0-102.0% ACS
Supplier: BeanTown Chemical
214070-100G
214070-500G
BT214070-100GEA
63.96
USD
BT214070-100G
BT214070-500G
Cobalt(II) chloride hexahydrate 98.0-102.0% ACS
Cobalt(II) chloride hexahydrate
CAS: 7791-13-1; EC No: 231-589-4; MDL No: MFCD00149652; RTECS: GG0200000
UN No: UN3077; Haz Class: 9; Packing Group: III
Crystalline; Linear Formula: CoCl2·6H2O; MW: 237.93
Hygroscopic
- RTECS: GG0200000
- Hygroscopic
- Crystalline
Formula:
CoCl₂·6H₂O MW: 237.93 g/mol |
MDL Number:
MFCD00149652 CAS Number: 7791-13-1 EINECS: 231-589-4 UN: 3077 ADR: 9,III |
Specification Test Results
Assay | 98.0-102.0% |
Insoluble matter | ≤0.01% |
Nitrate (NO3) | ≤0.01% |
Sulfate (SO4) | ≤0.01% |
Calcium (Ca) | ≤0.005% |
Copper (Cu) | ≤0.002% |
Iron (Fe) | ≤0.005% |
Magnesium (Mg) | ≤0.005% |
Nickel (Ni) | ≤0.1% |
Potassium (K) | ≤0.01% |
Sodium (Na) | ≤0.05% |
Zinc (Zn) | ≤0.03% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...