(S)-(-)-α-Methylbenzylamine ≥99%, ee 99% ee
Supplier: BeanTown Chemical
Synonyms:
(S)-(-)-1-Phenylethylamine, (S)-(-)-α-Phenylethylamine, L(-)-α-Methylbenzylamine
214065-100G
214065-25G
214065-5G
BT214065-100GEA
158.63
USD
BT214065-100G
BT214065-25G
BT214065-5G
(S)-(-)-α-Methylbenzylamine ≥99%, ee 99% ee
(S)-(-)-α-Methylbenzylamine
CAS: 2627-86-3; EC No: 220-098-0; MDL No: MFCD00064406
UN No: UN2735; Haz Class: 8; Packing Group: III
Liquid; Molecular Formula: C8H11N; MW: 121.18
Boiling Point: 187°; Flash point: 70°C (158°F)
Density (g/mL): 0.94; Refractive Index: 1.526; Optical Rotation: [α]20/D -39°, neat
Air Sensitive
- Optical rotation: [α]20/D -39°, neat
- Refractive index: 1.526
- Air Sensitive
- Liquid
Formula:
C₈H₁₁N MW: 121.18 g/mol Boiling Pt: 187 °C Density: 0.94 g/mL Flash Pt: 70 °C (158 °F) |
MDL Number:
MFCD00064406 CAS Number: 2627-86-3 EINECS: 220-098-0 ADR: 8,III |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...