Benzyl bromide ≥99%

Supplier: BeanTown Chemical

Synonyms: alpha-Bromotoluene

214050-1KG 214050-250G 214050-50G
BT214050-1KGEA 319.25 USD
BT214050-1KG BT214050-250G BT214050-50G
Benzyl bromide ≥99%
Benzyl bromide

CAS: 100-39-0; EC No: 202-847-3; MDL No: MFCD00000172; RTECS: XS7965000 UN No: UN1737; Haz Class: 6.1 (8); Packing Group: II Liquid; Linear Formula: C6H5CH2Br; Molecular Formula: C7H7Br; MW: 171.03 Melting Point: -3°; Boiling Point: 198-199°; Flash point: 86°C (187°F) Density (g/mL): 1.438; Refractive Index: 1.575 Moisture Sensitive


  • RTECS: XS7965000
  • Refractive index: 1.575
  • Moisture Sensitive
  • Liquid

Formula: (C₆H₅)CH₂Br
MW: 171.04 g/mol
Boiling Pt: 199
Melting Pt: -4
Density: 1.438 g/mL
Flash Pt: 86 °C (187 °F)
MDL Number: MFCD00000172
CAS Number: 100-39-0
EINECS: 202-847-3
UN: 1737
ADR: 6.1 II
Merck Index: 13,01130

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR