Sodium peroxide ≥93.0% ACS

Supplier: BeanTown Chemical

Synonyms: di-Sodium peroxide

214040-100G 214040-25G 214040-500G
BT214040-100GEA 113.62 USD
BT214040-100G BT214040-25G BT214040-500G
Sodium peroxide ≥93.0% ACS
Sodium peroxide

CAS: 1313-60-6; EC No: 215-209-4; MDL No: MFCD00003497; RTECS: WD3450000 UN No: UN1504; Haz Class: 5.1; Packing Group: I Powder; Molecular Formula: Na2O2 ; MW: 77.98 Melting Point: 460° (decomposes); Boiling Point: 657° Density (g/mL): 2.805 Air Sensitive , Hygroscopic


  • RTECS: WD3450000
  • Air Sensitive, Hygroscopic
  • Powder

Formula: Na₂O₂
Boiling Pt: 657 °C
Melting Pt: 460 °C (dec.)
Density: 2.805 g/mL
MDL Number: MFCD00003497
CAS Number: 1313-60-6
EINECS: 215-209-4
UN: 1504
ADR: 5.1,I

Order Now

Specification Test Results

Assay 93.0% min.
Chloride (Cl) ≤0.002%
Phosphate (PO4) ≤0.0005%
Sulfate (SO4) ≤0.001%
Heavy metals (as Pb) ≤0.002%
Iron (Fe) ≤0.005%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR