3,3-Dimethylbutyric acid ≥98%

Supplier: BeanTown Chemical

Synonyms: 3,3-Dimethylbutanoic acid, tert-Butylacetic acid

213995-100G 213995-25G
BT213995-100GEA 111 USD
BT213995-100G BT213995-25G
3,3-Dimethylbutyric acid ≥98%
3,3-Dimethylbutyric acid

CAS: 1070-83-3; EC No: 213-982-2; MDL No: MFCD00002715 UN No: UN3265; Haz Class: 8; Packing Group: III Liquid; Linear Formula: (CH3)3CCH2COOH; Molecular Formula: C6H11O2; MW: 116.16 Melting Point: -11°; Boiling Point: 185-190°; Flash point: 89°C (192°F) Density (g/mL): 0.912; Refractive Index: 1.411


  • Refractive index: 1.411
  • Liquid

Formula: C₆H₁₂O₂
MW: 116.16 g/mol
Boiling Pt: 185…190 °C
Melting Pt: –11 °C
Density: 0.912 g/mL
Flash Pt: 89 °C (192 °F)
MDL Number: MFCD00002715
CAS Number: 1070-83-3
EINECS: 213-982-2
UN: 3265
ADR: 8,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR