Tetrabutylammonium perchlorate ≥99%
Supplier: BeanTown Chemical
Synonyms:
Tetra-n-butylammonium perchlorate
213960-100G
213960-25G
213960-5G
BT213960-100GEA
423.67
USD
BT213960-100G
BT213960-25G
BT213960-5G
Tetrabutylammonium perchlorate ≥99%
Tetrabutylammonium perchlorate
CAS: 1923-70-2; EC No: 217-655-5; MDL No: MFCD00038722
UN No: UN1481; Haz Class: 5.1; Packing Group: II
Crystalline/Powder; Linear Formula: [CH3(CH2)3]4N(ClO4); Molecular Formula: C16H36ClNO4 ; MW: 341.92
Melting Point: 210-215°
Hygroscopic
- Hygroscopic
- Crystalline/Powder
Formula:
C₁₆H₃₆ClNO₄ MW: 341.92 g/mol Melting Pt: 210…215 °C |
MDL Number:
MFCD00038722 CAS Number: 1923-70-2 EINECS: 217-655-5 UN: 1481 ADR: 5.1,II |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...