Acrylamide ≥99%
Supplier: BeanTown Chemical
213920-100G
213920-25G
213920-500G
BT213920-100GEA
40.56
USD
BT213920-100G
BT213920-25G
BT213920-500G
Acrylamide ≥99%
Acrylamide
CAS: 79-06-1; EC No: 201-173-7; MDL No: MFCD00008032; RTECS: AS3325000
UN No: UN2074; Haz Class: 6.1; Packing Group: III
Crystalline; Linear Formula: CH2=CHCONH2; Molecular Formula: C3H5NO; MW: 71.08
Melting Point: 82-86°; Boiling Point: 125°/25 mmHg; Flash point: 138°C (280°F)
Light Sensitive
- RTECS: AS3325000
- Light Sensitive
- Crystalline
Formula:
C₃H₅NO Boiling Pt: 125 °C/25 mmHg Melting Pt: 82…86 °C Flash Pt: 138 °C (280 °F) |
MDL Number:
MFCD00008032 CAS Number: 79-06-1 EINECS: 201-173-7 UN: 2074 ADR: 6.1,III |
|
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...