Calcium hydride ≥90%, Technical Grade

Supplier: BeanTown Chemical

Synonyms: Calcium dihydride

213910-100G 213910-500G
BT213910-100GEA 101.54 USD
BT213910-100G BT213910-500G
Calcium hydride ≥90%, Technical Grade
Calcium hydride

CAS: 7789-78-8; EC No: 232-189-2; MDL No: MFCD00010897 UN No: UN1404; Haz Class: 4.3; Packing Group: I 1-20mm Granules; Molecular Formula: CaH2; MW: 42.09 Melting Point: 190° Moisture Sensitive


  • Moisture Sensitive
  • 1-20mm Granules

Formula: H₂Ca
MW: 42.09 g/mol
Melting Pt: 190 °C
MDL Number: MFCD00010897
CAS Number: 7789-78-8
EINECS: 232-189-2
UN: 1404
ADR: 4.3,I

Order Now

Specification Test Results

Assay 90%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR