Methylmagnesium chloride 3 M in THF
Supplier: BeanTown Chemical
213905-100ML
213905-50ML
BT213905-100MLEA
75.17
USD
BT213905-100ML
BT213905-50ML
Methylmagnesium chloride 3 M in THF
Methylmagnesium chloride
CAS: 676-58-4; EC No: 211-629-7; MDL No: MFCD00000468
UN No: UN3399; Haz Class: 4.3 (3); Packing Group: I
Liquid; Linear Formula: CH3MgCl; MW: 74.79
Flash point: -17°C (1°F)
Density (g/mL): 1.013
Air Sensitive, Moisture Sensitive
- Air Sensitive, Moisture Sensitive
- Liquid
Formula:
CH₃ClMg MW: 74.79 g/mol Density: 1.013 g/mL Flash Pt: –17 °C (1 °F) |
MDL Number:
MFCD00000468 CAS Number: 676-58-4 EINECS: 211-629-7 UN: 3399 ADR: 4.3,I |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...