Amidosulfonic acid 99.3-100.3% (dry weight) ACS
Supplier: BeanTown Chemical
Synonyms:
Aminosulphonic acid, Amidosulphuric acid, Amidosulphuric Acid, Sulphamic acid, Amidosulphonic acid, Sulphamidic acid
213870-100G
213870-500G
BT213870-100GEA
41.56
USD
BT213870-100G
BT213870-500G
Amidosulfonic acid 99.3-100.3% (dry weight) ACS
Amidosulphonic acid
CAS: 5329-14-6; EC No: 226-218-8; MDL No: MFCD00011603; RTECS: W05950000
UN No: UN2967; Haz Class: 8; Packing Group: III
Crystalline; Linear Formula: NH2SO3H; MW: 97.09
Melting Point: 215-225° (decomposes)
- RTECS: W05950000
- Crystalline
Formula:
H₃NO₃S MW: 97.09 g/mol Melting Pt: 215…225 °C (dec.) |
MDL Number:
MFCD00011603 CAS Number: 5329-14-6 EINECS: 226-218-8 UN: 2967 ADR: 8,III |
Specification Test Results
Assay | 99.3-100.3% |
Insoluble matter | ≤0.01% |
Residue after ignition | ≤0.01% |
Chloride (Cl) | ≤0.001% |
Sulfate (SO4) | ≤0.05% |
Heavy metals (as Pb) | ≤0.001% |
Iron (Fe) | ≤5ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...