Canada balsam

Supplier: BeanTown Chemical

213855-100G 213855-25G 213855-500G
BT213855-100GEA 103.04 USD
BT213855-100G BT213855-25G BT213855-500G
Canada balsam
Canada balsam

CAS: 8007-47-4; EC No: 232-362-2; MDL No: MFCD00132800; RTECS: CP2352500 UN No: UN1993; Haz Class: 3; Packing Group: III Liquid Flash point: 62°C (144°F) Density (g/mL): 0.99; Refractive Index: 1.522


  • RTECS: CP2352500
  • Refractive index: 1.522
  • Liquid

Density: 0.99 g/mL
Flash Pt: 62 °C (144 °F)
MDL Number: MFCD00132800
CAS Number: 8007-47-4
EINECS: 232-362-2
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR