Phenylphosphonic dichloride ≥90%, Technical Grade

Supplier: BeanTown Chemical

Synonyms: Phenyldichlorophosphine oxide, Dichlorophenylphosphine oxide

213780-1KG 213780-250G 213780-50G
BT213780-1KGEA 464 USD
BT213780-1KG BT213780-250G BT213780-50G
Phenylphosphonic dichloride ≥90%, Technical Grade
Phenylphosphonic dichloride

CAS: 824-72-6; EC No: 212-534-3; MDL No: MFCD00002070 UN No: UN3265; Haz Class: 8; Packing Group: II Liquid; Linear Formula: C6H5POCl2; MW: 194.98 Melting Point: 3°; Boiling Point: 258°; Flash point: 204°C (399°F) Density (g/mL): 1.375; Refractive Index: 1.559 Moisture Sensitive


  • Refractive index: 1.559
  • Moisture Sensitive
  • Liquid

Formula: C₆H₅Cl₂OP
MW: 194.98 g/mol
Boiling Pt: 258 °C
Melting Pt: 3 °C
Density: 1.375 g/mL
Flash Pt: 204 °C (399 °F)
MDL Number: MFCD00002070
CAS Number: 824-72-6
EINECS: 212-534-3
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Assay 90%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR