Pyridinium chlorochromate ≥98%
Supplier: BeanTown Chemical
Synonyms:
PCC
213775-100G
213775-25G
213775-500G
BT213775-100GEA
70.22
USD
BT213775-100G
BT213775-25G
BT213775-500G
Pyridinium chlorochromate ≥98%
Pyridinium chlorochromate
CAS: 26299-14-9; EC No: 247-595-5; MDL No: MFCD00013106
UN No: UN1479; Haz Class: 5.1; Packing Group: II
Crystalline/Powder; Linear Formula: C6H5N·CrClO3H; Molecular Formula: C5H6ClCrNO3; MW: 215.56
Melting Point: 205-208° (decomposes)
Moisture Sensitive
- Moisture Sensitive
- Crystalline/Powder
Formula:
C₅H₆ClCrNO₃ Melting Pt: 205…208 °C (dec.) |
MDL Number:
MFCD00013106 CAS Number: 26299-14-9 EINECS: 247-595-5 UN: 1479 ADR: 5.1,II |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...