4-Bromo-1-butanol, technical
Supplier: BeanTown Chemical
Synonyms:
1-Bromo-4-hydroxybutane
213755-1G
213755-25G
213755-5G
BT213755-1GEA
57
USD
BT213755-1G
BT213755-25G
BT213755-5G
4-Bromo-1-butanol, technical
4-Bromo-1-butanol
CAS: 33036-62-3; EC No: 251-355-5; MDL No: MFCD00015387
UN No: UN1987; Haz Class: 3; Packing Group: III
Liquid; Linear Formula: BrCH2CH2CH2CH2OH; Molecular Formula: C4H9BrO ; MW: 153.02
Flash point: 7°C (45°F)
Light Sensitive, Moisture Sensitive
- Light Sensitive, Moisture Sensitive
- Liquid
Formula:
BrH₂C(CH₂)₂CH₂OH MW: 153.02 Flash Pt: 7 °C (45 °F) |
MDL Number:
MFCD00015387 CAS Number: 33036-62-3 EINECS: 251-355-5 UN: 1987 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...