2-Chloro-5-(trifluoromethyl)nicotinic acid ≥98%
Supplier: BeanTown Chemical
213740-1G
213740-250MG
213740-5G
BT213740-1GEA
130
USD
BT213740-1G
BT213740-250MG
BT213740-5G
2-Chloro-5-(trifluoromethyl)nicotinic acid ≥98%
2-Chloro-5-(trifluoromethyl)nicotinic acid
CAS: 505084-59-3; MDL No: MFCD02257701
UN No: UN2811 ; Haz Class: 6.1; Packing Group: III
Solid; Molecular Formula: C7H3ClF3NO2; MW: 225.55
Melting Point: 173-178°
- Solid
Formula:
C₇H₃ClF₃NO₂ MW: 225.55 g/mol Melting Pt: 173…178 °C |
MDL Number:
MFCD08741353 CAS Number: 505084-59-3 ADR: 6.1,III |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...