1-benzyl-1,4-diazepane ≥98%
Supplier: BeanTown Chemical
Synonyms:
1-Benzylhomopiperazine, 1-Benzyl-1,4-diazepane
213715-1G
213715-5G
BT213715-1GEA
63
USD
BT213715-1G
BT213715-5G
1-benzyl-1,4-diazepane ≥98%
1-benzyl-1,4-diazepane
CAS: 4410-12-2; MDL No: MFCD00800858
UN No: UN2735; Haz Class: 8; Packing Group: III
Liquid; Molecular Formula: C12H18N2; MW: 190.28
Flash point: 110°C (230°F)
Density (g/mL): 1.028; Refractive Index: 1.547
Air Sensitive
- Refractive index: 1.547
- Air Sensitive
- Liquid
Formula:
C₁₂H₁₈N₂ MW: 190.29 g/mol Density: 1.028 g/mL Flash Pt: 110 °C (230 °F) |
MDL Number:
MFCD00800858 CAS Number: 4410-12-2 ADR: 8,III |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...