Thulium ≥99.9% (trace rare earth metals basis), foil, Thickness 0.1 mm

Supplier: BeanTown Chemical

213695-25X25MM 213695-50X50MM
BT213695-25X25EA 354 USD
BT213695-25X25 BT213695-50X50
Thulium ≥99.9% (trace rare earth metals basis), foil, Thickness 0.1 mm
Thulium

CAS: 7440-30-4; EC No: 231-140-2; MDL No: MFCD00011281 UN No: UN3178; Haz Class: 4.1; Packing Group: II Solid; Molecular Formula: Tm; MW: 168.93 Melting Point: 1545°; Boiling Point: 1950° Density (g/mL): 9.332


  • Solid

Formula: Tm
MW: 168.93 g/mol
Boiling Pt: 1950 °C
Melting Pt: 1545 °C
Density: 9.332 g/mL
MDL Number: MFCD00011281
CAS Number: 7440-30-4
EINECS: 231-140-2
UN: 3178
ADR: 4.1,II

Order Now

Specification Test Results

Assay 99.9%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR