Sodium perborate tetrahydrate

Supplier: BeanTown Chemical

Synonyms: Sodium peroxometaborate tetrahydrate

213570-100G 213570-25G 213570-500G
BT213570-100GEA 38.83 USD
BT213570-100G BT213570-25G BT213570-500G
Sodium perborate tetrahydrate
Sodium perborate tetrahydrate

CAS: 10486-00-7; EC No: 239-172-9; MDL No: MFCD00149231; RTECS: SC7350000 UN No: UN1479; Haz Class: 5.1; Packing Group: III Crystalline/Granular; Linear Formula: NaBO3·4H2O; MW: 153.86 Melting Point: 60° (decomposes) Density (g/mL): 1.73 Moisture Sensitive


  • RTECS: SC7350000
  • Moisture Sensitive
  • Crystalline/Granular

Formula: NaH₂BO₄·4H₂O
MW: 171.87 g/mol
Melting Pt: 60 (dec.)
Density: 1.73 g/mL
MDL Number: MFCD00149231
CAS Number: 13517-20-9
EINECS: 239-172-9
ADR: 5.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR