Tungsten(VI) chloride ≥99%

Supplier: BeanTown Chemical

Synonyms: Tungsten hexachloride

213540-10G 213540-50G
BT213540-10GEA 63 USD
BT213540-10G BT213540-50G
Tungsten(VI) chloride ≥99%
Tungsten(VI) chloride

CAS: 13283-01-7; EC No: 236-293-9; MDL No: MFCD00011463 UN No: UN3260; Haz Class: 8; Packing Group: II Crystalline; Linear Formula: WCl6; MW: 396.57 Melting Point: 275°; Boiling Point: 347° Density (g/mL): 3.52 Moisture Sensitive


  • Moisture Sensitive
  • Crystalline

Formula: Cl₆W
MW: 396.56 g/mol
Boiling Pt: 347
Melting Pt: 275
Density: 3.52 g/mL
MDL Number: MFCD00011463
CAS Number: 13283-01-7
EINECS: 236-293-9
UN: 3260
ADR: 8 II

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR