Aluminium acetylacetonate

Supplier: BeanTown Chemical

Synonyms: Tris(2,4-pentanedionato-O,O')aluminium, Acetylacetone Aluminum(III) Salt, Al(acac)3, Aluminium 2,4-pentanedionate, Aluminum(III) Acetylacetonate, Aluminium tris(2,4-pentanedionato-O,O'), Aluminum acetylacetonate, Aluminum 2,4-pentanedionate, Tris(2,4-pentanedionato)aluminum(III), Aluminum triacetylacetone

213520-100G 213520-500G
BT213520-100GEA 32 USD
BT213520-100G BT213520-500G
Aluminium acetylacetonate
Aluminium acetylacetonate

CAS: 13963-57-0; EC No: 237-741-6; MDL No: MFCD00000013; RTECS: BD2230000 UN No: UN3467; Haz Class: 6.1; Packing Group: II Powder; Molecular Formula: C15H21AlO6 ; MW: 324.31 Melting Point: 190-193°; Boiling Point: 315°


  • RTECS: BD2230000
  • Powder

Formula: C₁₅H₂₁AlO₆
MW: 324.31 g/mol
Boiling Pt: 315
Melting Pt: 190…193 °C
MDL Number: MFCD00000013
CAS Number: 13963-57-0
EINECS: 237-741-6
UN: 3467
ADR: 6.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR