Nickel plating solution, bright finish

Supplier: BeanTown Chemical

213510-1L 213510-250ML
BT213510-1LEA 67 USD
BT213510-1L BT213510-250ML
Nickel plating solution, bright finish
Nickel plating solution

MDL No: MFCD00011137 UN No: UN3082; Haz Class: 9; Packing Group: III Liquid Density (g/mL): 1.22


  • Liquid

Density: 1.22 g/mL
MDL Number: MFCD00011137
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR