Cyclopropyl methyl ketone ≥99%
Supplier: BeanTown Chemical
CAS: 765-43-5; EC No: 212-146-4; MDL No: MFCD00001297; RTECS: KM5648000
UN No: UN1992; Haz Class: 3 (6.1); Packing Group: II
Liquid; Molecular Formula: C5H8O ; MW: 84.12
Boiling Point: 114°; Flash point: 21°C (70°F)
Density (g/mL): 0.849; Refractive Index: 1.424
- RTECS: KM5648000
- Refractive index: 1.424
- Liquid
Formula:
C₅H₈O Boiling Pt: 114 °C Density: 0.849 g/mL Flash Pt: 21 °C (70 °F) |
MDL Number:
MFCD00001297 CAS Number: 765-43-5 EINECS: 212-146-4 UN: 1992 ADR: 3,II |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...