3,3-Dimethyl-1-butene ≥95%

Supplier: BeanTown Chemical

213405-250ML 213405-50ML
BT213405-250MLEA 77 USD
BT213405-250ML BT213405-50ML
3,3-Dimethyl-1-butene ≥95%
3,3-Dimethyl-1-butene

CAS: 558-37-2; EC No: 209-195-9; MDL No: MFCD00008853 UN No: UN3295; Haz Class: 3; Packing Group: II Liquid; Linear Formula: (CH3)3CCH=CH2; Molecular Formula: C6H12; MW: 84.16 Melting Point: -115°; Boiling Point: 41°; Flash point: -8°C (18°F) Density (g/mL): 0.653; Refractive Index: 1.376


  • Refractive index: 1.376
  • Liquid

Formula: C₆H₁₂
MW: 84.16 g/mol
Boiling Pt: 41 °C
Melting Pt: –115 °C
Density: 0.653 g/mL
Flash Pt: –8 °C (18 °F)
MDL Number: MFCD00008853
CAS Number: 558-37-2
EINECS: 209-195-9
UN: 3295
ADR: 3,II

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR