Coumarin ≥98%

Supplier: BeanTown Chemical

213400-1KG 213400-250G 213400-5KG
BT213400-1KGEA 159.88 USD
BT213400-1KG BT213400-250G BT213400-5KG
Coumarin ≥98%
Coumarin

CAS: 91-64-5; EC No: 202-086-7; MDL No: MFCD00006850; RTECS: GN4200000 UN No: UN2811; Haz Class: 6.1; Packing Group: III Flakes; Molecular Formula: C9H6O2 ; MW: 146.14 Melting Point: 68-73°; Boiling Point: 298°


  • RTECS: GN4200000
  • Flakes

Formula: C₉H₆O₂
Boiling Pt: 298 °C
Melting Pt: 68…73 °C
MDL Number: MFCD00006850
CAS Number: 91-64-5
EINECS: 202-086-7
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR