2,3-Dichloropyridine ≥99%

Supplier: BeanTown Chemical

213370-100G 213370-25G 213370-5G
BT213370-100GEA 855 USD
BT213370-100G BT213370-25G BT213370-5G
2,3-Dichloropyridine ≥99%
2,3-Dichloropyridine

CAS: 2402-77-9; EC No: 219-281-8; MDL No: MFCD00006229; RTECS: US8050000 UN No: UN2811; Haz Class: 6.1; Packing Group: III Crystalline; Molecular Formula: C5H3Cl2N; MW: 147.99 Melting Point: 64-67°


  • RTECS: US8050000
  • Crystalline

Formula: C₅H₃Cl₂N
MW: 147.99 g/mol
Melting Pt: 64…67 °C
MDL Number: MFCD00006229
CAS Number: 2402-77-9
EINECS: 219-281-8
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR