Magnesium perchlorate, anhydrous ACS

Supplier: BeanTown Chemical

Synonyms: Anhydrone

213360-100G 213360-500G
BT213360-100GEA 70.22 USD
BT213360-100G BT213360-500G
Magnesium perchlorate, anhydrous ACS
Magnesium perchlorate

CAS: 10034-81-8; EC No: 233-108-3; MDL No: MFCD00011102 UN No: UN1475; Haz Class: 5.1; Packing Group: II Granular; Linear Formula: Mg(ClO4)2; MW: 223.20 Melting Point: 251° (decomposes) Density (g/mL): 2.21 Hygroscopic


  • Hygroscopic
  • Granular

Formula: Cl₂MgO₈
MW: 223.21 g/mol
Melting Pt: 251 °C (dec.)
Density: 2.21 g/mL
MDL Number: MFCD00011102
CAS Number: 10034-81-8
EINECS: 233-108-3
UN: 1475
ADR: 5.1,II

Order Now

Specification Test Results

Water ≤8%
Titrable free acid ≤0.005meq/g
Titrable base ≤0.025meq/g
Suitability for moisture absorption Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR