4-Fluoro-2-nitrotoluene ≥98%
Supplier: BeanTown Chemical
213350-100G
213350-25G
213350-5G
BT213350-100GEA
373
USD
BT213350-100G
BT213350-25G
BT213350-5G
4-Fluoro-2-nitrotoluene ≥98%
4-Fluoro-2-nitrotoluene
CAS: 446-10-6; EC No: 207-161-8; MDL No: MFCD00007200
UN No: UN2811; Haz Class: 6.1; Packing Group: III
Liquid/Low Melting Solid; Molecular Formula: C7H6FNO2 ; MW: 155.13
Melting Point: 27°; Boiling Point: 138-139°/83 mmHg; Flash point: 99°C (210°F)
Density (g/mL): 1.26; Refractive Index: 1.522
- Refractive index: 1.522
- Liquid/Low Melting Solid
Formula:
C₇H₆FNO₂ MW: 155.13 g/mol Boiling Pt: 138…139 °C/83 mmHg Melting Pt: 27 °C Density: 1.26 g/mL Flash Pt: 99 °C (210 °F) |
MDL Number:
MFCD00007200 CAS Number: 446-10-6 EINECS: 207-161-8 UN: 2811 ADR: 6.1,III |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...